Gene Transcripts

Gene Symbol
SPX
Entrez Gene ID
80763
Gene Name
spexin hormone
Gene Aliases
C12orf39
Chromosome Location
Chr. 12: 21526257 - 21532947 on Build GRCh38
UniGene ID
Hs.130692
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030572.3 NP_085049.1 3 342
GenBank mRNA AK027273.1 - 3 180
AK075342.1 - 3 296
BC004336.1 - 3 277 3631700
DA380664.1 - 3 341

View Full Product Details