Gene Transcripts

Gene Symbol
SRXN1
Entrez Gene ID
140809
Gene Name
sulfiredoxin 1
Gene Aliases
C20orf139, Npn3, SRX, SRX1
Chromosome Location
Chr. 20: 646615 - 653370 on Build GRCh38
UniGene ID
Hs.516830
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080725.2 NP_542763.1 2 430
GenBank mRNA AK127435.1 - 2 275
AK312054.1 - 2 260
AL833944.1 - 2 306
BC017001.1 - 2 225
BC032604.1 - 2 261 5535883
BC047707.1 - 2 250 5735551
BU170568.1 - 395

View Full Product Details