Gene Transcripts

Gene Symbol
HSCB
Entrez Gene ID
150274
Gene Name
HscB mitochondrial iron-sulfur cluster cochaperone
Gene Aliases
DNAJC20, HSC20, JAC1
Chromosome Location
Chr. 22: 28741992 - 28757518 on Build GRCh38
UniGene ID
Hs.632780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318316.1 NP_001305245.1 4 374
NM_172002.4 NP_741999.3 4 620
NR_134560.1 4 565
XM_005261359.3 XP_005261416.1 4 621
XM_017028619.1 XP_016884108.1 4 591
XM_017028620.1 XP_016884109.1 4 424
GenBank mRNA AY191719.1 - 4 570
BC000004.1 - 4 629
BC065569.1 - 4 593 6171959
CR456462.1 - 4 588
HY057929.1 - 4 374

View Full Product Details