Gene Transcripts

Gene Symbol
FBXL19
Entrez Gene ID
54620
Gene Name
F-box and leucine rich repeat protein 19
Gene Aliases
CXXC11, Fbl19, JHDM1C
Chromosome Location
Chr. 16: 30923055 - 30948783 on Build GRCh38
UniGene ID
Hs.152149
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099784.2 NP_001093254.2 11 2660
NM_001282351.1 NP_001269280.1 11 2528
GenBank mRNA AK127701.1 - 11 2512
AL137589.1 - 3 870
BC059173.1 - 10 2176 5738074

View Full Product Details