Gene Transcripts

Gene Symbol
FAM13C
Entrez Gene ID
220965
Gene Name
family with sequence similarity 13 member C
Gene Aliases
FAM13C1
Chromosome Location
Chr. 10: 59245933 - 59363186 on Build GRCh38
UniGene ID
Hs.607594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001971.2 NP_001001971.1 10 1308
NM_001143773.1 NP_001137245.1 13 1773
NM_001166698.1 NP_001160170.1 13 1684
NM_198215.3 NP_937858.2 12 1602
XM_005269618.4 XP_005269675.2 12 1599
XM_005269619.4 XP_005269676.2 10 1305
XM_006717702.3 XP_006717765.1 14 1603
XM_006717703.3 XP_006717766.1 14 1785
XM_011539490.2 XP_011537792.1 13 1658
XM_011539491.2 XP_011537793.1 14 1704
XM_017015885.1 XP_016871374.1 13 1516
XM_017015886.1 XP_016871375.1 13 1770
XM_017015887.1 XP_016871376.1 13 1520
XM_017015888.1 XP_016871377.1 14 1782
XM_017015889.1 XP_016871378.1 14 1600
XM_017015890.1 XP_016871379.1 14 1839
XM_017015891.1 XP_016871380.1 11 1479
XM_017015892.1 XP_016871381.1 11 1476
XM_017015893.1 XP_016871382.1 13 1580
GenBank mRNA AK289796.1 - 12 1528
AK294805.1 - 12 1517
AK299924.1 - 13 1773
AK316542.1 - 13 1680
BC036453.1 - 12 1560 5270033
BC064431.1 - 10 1308 5764753

View Full Product Details