Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001258017.1 | NP_001244946.1 | 7 | 1023 | |
NM_001258018.1 | NP_001244947.1 | 7 | 1105 | ||
NM_152899.1 | NP_690863.1 | 5 | 496 | ||
NM_172374.2 | NP_758962.1 | 7 | 1047 | ||
NR_047577.1 | 8 | 1082 | |||
GenBank mRNA | AF293462.1 | - | 5 | 496 | |
AJ880386.1 | - | 7 | 922 | ||
AJ880387.1 | - | 7 | 899 | ||
AJ880388.1 | - | 7 | 961 | ||
AJ880389.1 | - | 8 | 959 | ||
AY358933.1 | - | 5 | 496 | ||
BC026103.1 | - | 7 | 951 | ||
BC064378.1 | - | 5 | 482 | 5725335 | |
BC090852.1 | - | 5 | 483 | 6191892 | |
BC131625.1 | - | 7 | 936 | 40117891 | |
DQ079587.1 | - | 7 | 1047 | ||
DQ079588.1 | - | 7 | 1105 | ||
DQ079589.1 | - | 7 | 1023 |