Gene Transcripts

Gene Symbol
OR52E6
Entrez Gene ID
390078
Gene Name
olfactory receptor family 52 subfamily E member 6
Gene Aliases
OR11-58
Chromosome Location
Chr. 11: 5840956 - 5841897 on Build GRCh38
UniGene ID
Hs.534693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005167.1 NP_001005167.1 1 895

View Full Product Details