Gene Transcripts

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
Chromosome Location
Chr. 2: 110637698 - 110678107 on Build GRCh38
UniGene ID
Hs.469649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278616.1 NP_001265545.1 22 2936
NM_001278617.1 NP_001265546.1 22 2825
NM_004336.4 NP_004327.1 23 2996
GenBank mRNA AF011387.1 - 15 2061
AF043294.2 - 23 2916
AF046078.1 - 23 2933
AF047471.1 - 23 2919
AF053305.1 - 23 2947
AK225317.1 - 22 2781
AK291221.1 - 23 2949
AK302418.1 - 22 2892
BC028201.1 - 23 2917 5205385

View Full Product Details