Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142805.1 | NP_001136277.1 | 9 | 1565 | |
NM_001142806.1 | NP_001136278.1 | 9 | 1144 | ||
NM_005629.3 | NP_005620.1 | 9 | 1595 | ||
GenBank mRNA | AB209704.1 | - | 9 | 1369 | |
AK295495.1 | - | 9 | 1144 | ||
BC012355.1 | - | 9 | 1362 | 4634609 | |
BC081558.1 | - | 9 | 1596 | 4823748 | |
EU280316.1 | - | 5 | 1356 | ||
KJ534943.1 | - | 9 | 1317 | ||
KJ534944.1 | - | 9 | 1317 | ||
KJ534945.1 | - | 6 | 876 | ||
L31409.1 | - | 9 | 1790 | ||
U17986.1 | - | 4 | 1475 |