Gene Transcripts
- Gene Symbol
- SGSH
- Entrez Gene ID
- 6448
- Gene Name
- N-sulfoglucosamine sulfohydrolase
- Gene Aliases
- HSS, MPS3A, SFMD
- Chromosome Location
- Chr. 17: 80200668 - 80220400 on Build GRCh38
- UniGene ID
- Hs.31074
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000199.3 |
NP_000190.1
|
5, 6 |
748 |
|
XM_005257582.3 |
XP_005257639.1
|
5, 6 |
681 |
|
XM_005257583.4 |
XP_005257640.1
|
5, 6 |
681 |
|
XM_011525126.1 |
XP_011523428.1
|
5, 6 |
681 |
|
XM_011525127.2 |
XP_011523429.1
|
5, 6 |
681 |
|
XM_017024951.1 |
XP_016880440.1
|
5, 6 |
681 |
|
XM_017024952.1 |
XP_016880441.1
|
5, 6 |
681 |
|
GenBank mRNA |
AK095969.1 |
- |
4, 5 |
602 |
|
AK222890.1 |
- |
5, 6 |
681 |
|
AK291257.1 |
- |
5, 6 |
681 |
|
AK304706.1 |
- |
5, 6 |
720 |
|
BC047318.1 |
- |
5, 6 |
673 |
5226903 |
U30894.1 |
- |
5, 6 |
673 |
|
View Full Product Details