Gene Transcripts

Gene Symbol
LRRN1
Entrez Gene ID
57633
Gene Name
leucine rich repeat neuronal 1
Gene Aliases
FIGLER3, NLRR-1
Chromosome Location
Chr. 3: 3799431 - 3849834 on Build GRCh38
UniGene ID
Hs.163244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324188.1 NP_001311117.1 2 747
NM_001324189.1 NP_001311118.1 2 670

View Full Product Details