Gene Transcripts
- Gene Symbol
- GPR89A
- Entrez Gene ID
- 653519
- Gene Name
- G protein-coupled receptor 89A
- Gene Aliases
- GPHR, GPR89, GPR89B, SH120, UNQ192
- Chromosome Location
- Chr. 1: 145607960 - 145670977 on Build GRCh38
- UniGene ID
- Hs.504320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001097612.1 |
NP_001091081.1
|
12, 13 |
1267 |
|
NM_001097613.2 |
NP_001091082.2
|
13, 14 |
1313 |
|
NR_036541.1 |
|
12, 13 |
1287 |
|
XM_006711492.3 |
XP_006711555.1
|
12, 13 |
1237 |
|
XM_006711493.3 |
XP_006711556.1
|
13, 14 |
1245 |
|
XM_011509908.2 |
XP_011508210.1
|
13, 14 |
1283 |
|
XM_011509909.2 |
XP_011508211.1
|
12, 13 |
1495 |
|
GenBank mRNA |
AB097024.1 |
- |
12, 13 |
1208 |
|
AB097025.1 |
- |
12, 13 |
1208 |
|
AF132947.1 |
- |
12, 13 |
1192 |
|
AK001354.1 |
- |
5, 6 |
2005 |
|
AK021758.1 |
- |
12, 13 |
1257 |
|
AK094228.1 |
- |
3, 4 |
1208 |
|
AK223223.1 |
- |
12, 13 |
1214 |
|
AK301939.1 |
- |
12, 13 |
1114 |
|
AK316371.1 |
- |
13, 14 |
1225 |
|
AY358966.1 |
- |
12, 13 |
1242 |
|
BC003187.1 |
- |
12, 13 |
1170 |
3537949 |
BC067816.1 |
- |
12, 13 |
1226 |
5295014 |
BC146880.1 |
- |
12, 13 |
1188 |
9056808 |
BT006679.1 |
- |
12, 13 |
1093 |
|
KJ534847.1 |
- |
12, 13 |
1093 |
|
KJ534848.1 |
- |
12, 13 |
1099 |
|
KJ535029.1 |
- |
11, 12 |
1093 |
|
KJ535049.1 |
- |
12, 13 |
1113 |
|
- Gene Symbol
- GPR89B
- Entrez Gene ID
- 51463
- Gene Name
- G protein-coupled receptor 89B
- Gene Aliases
- GPHR, GPR89, GPR89C, SH120, UNQ192
- Chromosome Location
- Chr. 1: 147928393 - 147993523 on Build GRCh38
- UniGene ID
- Hs.504320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016334.3 |
NP_057418.1
|
12, 13 |
1237 |
|
XM_005277401.3 |
XP_005277458.1
|
12, 13 |
1210 |
|
XM_005277402.4 |
XP_005277459.1
|
12, 13 |
1374 |
|
XM_005277404.3 |
XP_005277461.1
|
12, 13 |
1230 |
|
XM_006711377.3 |
XP_006711440.1
|
13, 14 |
1273 |
|
XM_006711378.2 |
XP_006711441.1
|
13, 14 |
1276 |
|
XM_011509610.2 |
XP_011507912.1
|
13, 14 |
1270 |
|
XM_011509612.2 |
XP_011507914.1
|
13, 14 |
1231 |
|
XM_017001448.1 |
XP_016856937.1
|
13, 14 |
1773 |
|
GenBank mRNA |
AF151035.1 |
- |
4, 5 |
349 |
|
AK299995.1 |
- |
12, 13 |
1208 |
|
AK302266.1 |
- |
12, 13 |
1229 |
|
AL133020.1 |
- |
12, 13 |
1738 |
|
CD102528.1 |
- |
7, 8 |
602 |
|
KJ534849.1 |
- |
12, 13 |
1099 |
|
KJ534850.1 |
- |
12, 13 |
1093 |
|
KJ534851.1 |
- |
12, 13 |
1024 |
|
KJ535070.1 |
- |
13, 14 |
1421 |
|
U78723.1 |
- |
12, 13 |
1196 |
|
View Full Product Details