Gene Transcripts

Gene Symbol
Esx1
Entrez Gene ID
13984
Gene Name
extraembryonic, spermatogenesis, homeobox 1
Gene Aliases
Spx1
Chromosome Location
Chr. X: 137115397 - 137122131 on Build GRCm38
UniGene ID
Mm.8017
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007957.2 NP_031983.2 3 625
GenBank mRNA AF004211.1 - 2 713
AF017734.1 - 2 342
AF017735.1 - 3 425
AF085715.1 - 3 626

View Full Product Details