Gene Transcripts

Gene Symbol
Lcn4
Entrez Gene ID
16821
Gene Name
lipocalin 4
Gene Aliases
A630045M08Rik, Vnsp2
Chromosome Location
Chr. 2: 26667674 - 26671282 on Build GRCm38
UniGene ID
Mm.123925
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010695.1 NP_034825.1 5, 6 530
GenBank mRNA BC114544.1 - 5, 6 529
D38581.1 - 5, 6 530

View Full Product Details