Gene Transcripts

Gene Symbol
Rnf13
Entrez Gene ID
24017
Gene Name
ring finger protein 13
Gene Aliases
2010001H16Rik, Rzf
Chromosome Location
Chr. 3: 57736043 - 57835233 on Build GRCm38
UniGene ID
Mm.274360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF037205.1 - 4 318
AF037206.1 - 4 318
AI882163.1 - 4 377
AK034135.1 - 4 369
AK036591.1 - 4 363
AK050095.1 - 4 331
AK082296.1 - 4 359
AK139447.1 - 4 324
AK140682.1 - 4 373
AK158046.1 - 4 359
AK167278.1 - 4 323
BB847350.1 - 4 384
BC058182.1 - 4 326 5324040

View Full Product Details