Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172809.3 | NP_766397.2 | 8 | 1425 | |
GenBank mRNA | AK038655.1 | - | 2 | 989 | |
AK046501.1 | - | 7 | 1280 | ||
AK053626.1 | - | 7 | 859 | ||
AK160797.1 | - | 8 | 1426 | ||
BC138482.1 | - | 7 | 981 | 8861502 | |
BC138483.1 | - | 7 | 981 | 8861503 | |
BC171956.1 | - | 7 | 981 | 9054242 |