Gene Transcripts

Gene Symbol
Mtmr7
Entrez Gene ID
54384
Gene Name
myotubularin related protein 7
Gene Aliases
Chromosome Location
Chr. 8: 40550159 - 40634792 on Build GRCm38
UniGene ID
Mm.294871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040699.1 NP_001035789.1 12 1553
GenBank mRNA AF073882.1 - 5 612
AK028160.1 - 12 1519
AK081973.1 - 12 1539
AK148768.1 - 12 1554
BC032254.1 - 13 1643 5368816
BC038468.1 - 2 220

View Full Product Details