Gene Transcripts

Gene Symbol
Pidd1
Entrez Gene ID
57913
Gene Name
p53 induced death domain protein 1
Gene Aliases
1200011D09Rik, AU042446, Lrdd, Pidd
Chromosome Location
Chr. 7: 141438113 - 141444408 on Build GRCm38
UniGene ID
Mm.334321
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022654.1 NP_073145.1 7 1438
GenBank mRNA AF274973.1 - 7 1438
AK004702.1 - 8 1381
AK156863.1 - 9 1426
AK183598.1 - 3 185
BC032202.1 - 7 1399
BC056987.1 - 7 1789
BC089373.1 - 7 1410
BC099686.1 - 7 1776
BC145314.1 - 7 1442
BC145315.1 - 7 1403
BC145857.1 - 7 1403 40131160

View Full Product Details