Gene Transcripts
- Gene Symbol
- Lamtor1
- Entrez Gene ID
- 66508
- Gene Name
- late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
- Gene Aliases
- 2400001E08Rik, Pdro, p18
- Chromosome Location
- Chr. 7: 101905837 - 101911903 on Build GRCm38
- UniGene ID
- Mm.214841
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013389.1 |
- |
3 |
308 |
|
View Full Product Details