Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001606.4 | NP_001597.2 | 6 | 661 | |
NM_212533.2 | NP_997698.1 | 6 | 653 | ||
XM_006716996.3 | XP_006717059.1 | 5 | 595 | ||
GenBank mRNA | AF178941.1 | - | 5 | 563 | |
AF327657.1 | - | 5 | 519 | ||
AK308738.1 | - | 6 | 558 | ||
BC029282.1 | - | 5 | 619 | ||
BC064542.1 | - | 6 | 549 | ||
BC144588.1 | - | 5 | 643 | ||
BM921499.1 | - | 5 | 609 | ||
CD676110.1 | - | 6 | 599 |