Gene Transcripts

Gene Symbol
Ndnf
Entrez Gene ID
68169
Gene Name
neuron-derived neurotrophic factor
Gene Aliases
A930038C07Rik, epidermacan
Chromosome Location
Chr. 6: 65671611 - 65712329 on Build GRCm38
UniGene ID
Mm.211177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172399.3 NP_765987.2 4 1425
GenBank mRNA AK029210.1 - 4 1426
AK049802.1 - 4 1450
BC047154.1 - 4 1399 6489681

View Full Product Details