Gene Transcripts

Gene Symbol
Ncapd2
Entrez Gene ID
68298
Gene Name
non-SMC condensin I complex, subunit D2
Gene Aliases
2810406C15Rik, 2810465G24Rik, CAP-D2, CNAP1, mKIAA0159
Chromosome Location
Chr. 6: 125168007 - 125191668 on Build GRCm38
UniGene ID
Mm.257590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146171.1 NP_666283.1 26, 27 3560
GenBank mRNA AK019259.2 - 12, 13 1724
AK129068.1 - 26, 27 3525
AK156143.1 - 26, 27 3590
BC006673.1 - 19, 20 2706
BC008592.1 - 17, 18 2454
BC023750.1 - 12, 13 1589
BC025460.1 - 26, 27 3580
BC026982.1 - 26, 27 3560 5364933
BC029133.1 - 26, 27 3517 4951596

View Full Product Details