Gene Transcripts

Gene Symbol
Lyzl6
Entrez Gene ID
69444
Gene Name
lysozyme-like 6
Gene Aliases
1700023H08Rik, Lyc1
Chromosome Location
Chr. 11: 103631069 - 103638889 on Build GRCm38
UniGene ID
Mm.275677
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027083.1 NP_081359.1 4, 5 568
GenBank mRNA AK006279.1 - 4, 5 485
AY566284.1 - 3, 4 420
BC048617.1 - 4, 5 568 6742605

View Full Product Details