Gene Transcripts

Gene Symbol
Mettl23
Entrez Gene ID
74319
Gene Name
methyltransferase like 23
Gene Aliases
1110005A03Rik, 1500035B17Rik, 4933424L15Rik
Chromosome Location
Chr. 11: 116843515 - 116849740 on Build GRCm38
UniGene ID
Mm.429312
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028865.3 NP_083141.3 5 722
GenBank mRNA AK003450.1 - 5 568
AK135294.1 - 849
AK160133.1 - 4 642
BC026936.1 - 5 637 4021089

View Full Product Details