Gene Transcripts

Gene Symbol
Snx7
Entrez Gene ID
76561
Gene Name
sorting nexin 7
Gene Aliases
2510028H01Rik
Chromosome Location
Chr. 3: 117781497 - 117868936 on Build GRCm38
UniGene ID
Mm.33824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029655.3 NP_083931.2 8 1219
GenBank mRNA AK011015.1 - 8 1194
AK038971.1 - 8 1220
AK052487.1 - 8 1176
BC087942.1 - 8 1077 30303462

View Full Product Details