Gene Transcripts

Gene Symbol
Ltn1
Entrez Gene ID
78913
Gene Name
listerin E3 ubiquitin protein ligase 1
Gene Aliases
4930528H02Rik, AV266914, C87237, Listerin, Rnf160, Zfp294
Chromosome Location
Chr. 16: 87376651 - 87432619 on Build GRCm38
UniGene ID
Mm.249005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081068.1 NP_001074537.1 26 4738
GenBank mRNA AK173009.1 - 26 4609
BC027795.1 - 5 708
BC030355.1 - 1 79

View Full Product Details