Gene Transcripts

Gene Symbol
Mapre3
Entrez Gene ID
100732
Gene Name
microtubule-associated protein, RP/EB family, member 3
Gene Aliases
AI790651, EB2, EB3
Chromosome Location
Chr. 5: 30814614 - 30866106 on Build GRCm38
UniGene ID
Mm.332295
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133350.1 NP_579928.1 6 825
GenBank mRNA AF468005.1 - 6 825
AK214734.1 - 1 57
BC048833.1 - 5 1387
BC057918.1 - 6 825 5134461
U51204.1 - 4 507

View Full Product Details