Gene Transcripts
- Gene Symbol
- JARID2-AS1
- Entrez Gene ID
- 100506681
- Gene Name
- JARID2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 15247815 - 15248696 on Build GRCh38
- UniGene ID
- Hs.664092
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_120502.1 |
|
2 |
252 |
|
GenBank mRNA |
BC033996.1 |
- |
2 |
213 |
|
HY051522.1 |
- |
2 |
253 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY043093.1 |
- |
2 |
226 |
|
Chr. 6 - 15247832 - 15248669 [-] |
GenBankmRNA |
BI561918.1 |
- |
2 |
216 |
|
Chr. 6 - 15247815 - 15248656 [-] |
GENCODE |
ENST00000441978.1 |
- |
2 |
190 |
|
Chr. 6 - 15247815 - 15248634 [-] |
View Full Product Details