Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153788.3 | NP_722483.2 | 6 | 614 | |
GenBank mRNA | AK133473.1 | - | 4 | 1089 | |
AK138736.1 | - | 6 | 606 | ||
AK154210.1 | - | 6 | 579 | ||
AK154449.1 | - | 6 | 617 | ||
AK155001.1 | - | 6 | 635 | ||
BC031462.1 | - | 6 | 614 | 4017986 | |
BC037481.1 | - | 1 | 241 | 4460639 |