Gene Transcripts

Gene Symbol
Pld4
Entrez Gene ID
104759
Gene Name
phospholipase D family, member 4
Gene Aliases
AI132321, thss
Chromosome Location
Chr. 12: 112760655 - 112768988 on Build GRCm38
UniGene ID
Mm.203915
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178911.4 NP_849242.1 5 613
GenBank mRNA AK045459.1 - 5 572
AK079549.1 - 5 572
AK079878.1 - 5 615
AK151551.1 - 5 847
AK152969.1 - 4 460
AK170362.1 - 5 570
BC058565.1 - 5 563 6442038

View Full Product Details