Gene Transcripts

Gene Symbol
Vmn1r75
Entrez Gene ID
171241
Gene Name
vomeronasal 1 receptor 75
Gene Aliases
V1rg6
Chromosome Location
Chr. 7: 11880343 - 11881260 on Build GRCm38
UniGene ID
Mm.377197
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134207.1 NP_598968.1 1 196
GenBank mRNA BC115506.1 - 1 189
BC115507.1 - 1 196 40101051

View Full Product Details