Gene Transcripts

Gene Symbol
Armc7
Entrez Gene ID
276905
Gene Name
armadillo repeat containing 7
Gene Aliases
A630084C13, BC023126
Chromosome Location
Chr. 11: 115475638 - 115490468 on Build GRCm38
UniGene ID
Mm.38019
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177778.4 NP_808446.1 1, 2 359
GenBank mRNA AK042345.1 - 1, 2 195
BC023126.1 - 1, 2 359 2616092

View Full Product Details