Gene Transcripts

Gene Symbol
Spink10
Entrez Gene ID
328971
Gene Name
serine peptidase inhibitor, Kazal type 10
Gene Aliases
A230091H23Rik
Chromosome Location
Chr. 18: 62548911 - 62662933 on Build GRCm38
UniGene ID
Mm.312675
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177829.3 NP_808497.1 4, 5 335
GenBank mRNA AK039062.1 - 4, 5 336
AK162840.1 - 4, 5 303
BC125360.1 - 4, 5 251 40129875
BC125362.1 - 4, 5 251 40129877
DQ437330.1 - 4, 5 335

View Full Product Details