Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001006658.2 | NP_001006659.1 | 4 | 802 | |
NM_001877.4 | NP_001868.2 | 4 | 802 | ||
XM_011509206.2 | XP_011507508.1 | 4 | 698 | ||
GenBank mRNA | AK223627.1 | - | 4 | 774 | |
AK301496.1 | - | 4 | 770 | ||
BC090937.1 | - | 4 | 737 | 3066233 | |
BC136394.1 | - | 4 | 776 | 9020381 | |
J03565.1 | - | 4 | 746 | ||
M26004.1 | - | 4 | 752 | ||
Y00649.1 | - | 4 | 738 |