Gene Transcripts
- Gene Symbol
- AMHR2
- Entrez Gene ID
- 269
- Gene Name
- anti-Mullerian hormone receptor type 2
- Gene Aliases
- AMHR, MISR2, MISRII, MRII
- Chromosome Location
- Chr. 12: 53423855 - 53431672 on Build GRCh38
- UniGene ID
- Hs.659889
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164690.1 |
NP_001158162.1
|
2, 3 |
307 |
|
NM_001164691.1 |
NP_001158163.1
|
2, 3 |
307 |
|
NM_020547.2 |
NP_065434.1
|
2, 3 |
307 |
|
XM_011538173.1 |
XP_011536475.1
|
2, 3 |
307 |
|
XM_011538174.1 |
XP_011536476.1
|
2, 3 |
307 |
|
XM_011538176.1 |
XP_011536478.1
|
2, 3 |
307 |
|
XM_011538178.1 |
XP_011536480.1
|
2, 3 |
307 |
|
XM_011538179.1 |
XP_011536481.1
|
2, 3 |
307 |
|
XM_011538180.1 |
XP_011536482.1
|
2, 3 |
307 |
|
XM_011538181.1 |
XP_011536483.1
|
2, 3 |
307 |
|
XM_011538183.2 |
XP_011536485.1
|
2, 3 |
307 |
|
XM_011538184.2 |
XP_011536486.1
|
2, 3 |
307 |
|
XM_011538185.1 |
XP_011536487.1
|
2, 3 |
307 |
|
XM_011538187.2 |
XP_011536489.1
|
2, 3 |
307 |
|
XM_017019179.1 |
XP_016874668.1
|
2, 3 |
307 |
|
GenBank mRNA |
AF172932.1 |
- |
2, 3 |
290 |
|
AK313593.1 |
- |
2, 3 |
296 |
|
AY714878.1 |
- |
2, 3 |
227 |
|
BC126316.1 |
- |
2, 3 |
282 |
8992032 |
BC136356.1 |
- |
2, 3 |
282 |
9020343 |
DB020573.1 |
- |
2, 3 |
308 |
|
View Full Product Details