Gene Transcripts

Gene Symbol
CASP3
Entrez Gene ID
836
Gene Name
caspase 3
Gene Aliases
CPP32, CPP32B, SCA-1
Chromosome Location
Chr. 4: 184627696 - 184649475 on Build GRCh38
UniGene ID
Hs.141125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004346.3 NP_004337.2 4 342
NM_032991.2 NP_116786.1 3 175
XM_011532301.1 XP_011530603.1 4 276
GenBank mRNA AJ413269.1 - 2 79
AK291337.1 - 4 313
AU127028.1 - 3 175
AU133051.1 - 4 314
BC016926.2 - 4 294 4419175
BX647609.1 - 4 414
U13737.1 - 4 303
U13738.1 - 3 142
U26943.1 - 2 79

View Full Product Details