Gene Transcripts
- Gene Symbol
- PON1
- Entrez Gene ID
- 5444
- Gene Name
- paraoxonase 1
- Gene Aliases
- ESA, MVCD5, PON
- Chromosome Location
- Chr. 7: 95298357 - 95324572 on Build GRCh38
- UniGene ID
- Hs.370995
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
D84371.1 |
- |
9 |
2228 |
|
U53784.1 |
- |
9 |
2228 |
|
Z70723.1 |
- |
9 |
2228 |
|
View Full Product Details