Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005898.4 | NP_005889.3 | 7 | 899 | |
NM_203364.2 | NP_976240.1 | 7 | 899 | ||
XM_017017749.1 | XP_016873238.1 | 6 | 870 | ||
GenBank mRNA | AK127972.1 | - | 6 | 699 | |
BC001731.1 | - | 7 | 899 | 3355481 | |
Z48042.1 | - | 6 | 752 |