Gene Transcripts

Gene Symbol
NEUROD6
Entrez Gene ID
63974
Gene Name
neuronal differentiation 6
Gene Aliases
Atoh2, MATH2, Math-2, NEX1M, Nex1, bHLHa2
Chromosome Location
Chr. 7: 31337461 - 31340924 on Build GRCh38
UniGene ID
Hs.45152
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022728.3 NP_073565.2 2 863
GenBank mRNA AF063609.1 - 2 666
AF136169.1 - 2 631
AF248954.1 - 2 863
AK055224.1 - 1 1960
AK055238.1 - 2 833
AK289619.1 - 2 664
BC035048.2 - 2 665 5261681

View Full Product Details