Gene Transcripts
- Gene Symbol
- ADAM28
- Entrez Gene ID
- 10863
- Gene Name
- ADAM metallopeptidase domain 28
- Gene Aliases
- ADAM 28, MDC-L, MDCL, eMDC II, eMDCII
- Chromosome Location
- Chr. 8: 24294040 - 24359018 on Build GRCh38
- UniGene ID
- Hs.174030
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304351.1 |
NP_001291280.1
|
11, 12 |
1214 |
|
NM_014265.5 |
NP_055080.2
|
11, 12 |
1214 |
|
NM_021777.4 |
NP_068547.2
|
11, 12 |
1214 |
|
NR_130709.1 |
|
10, 11 |
1021 |
|
NR_130710.1 |
|
11, 12 |
1276 |
|
XM_005273380.3 |
XP_005273437.1
|
11, 12 |
1214 |
|
XM_005273382.1 |
XP_005273439.1
|
11, 12 |
1214 |
|
XM_006716273.3 |
XP_006716336.1
|
11, 12 |
1214 |
|
XM_006716274.1 |
XP_006716337.1
|
11, 12 |
1214 |
|
XM_011544367.2 |
XP_011542669.1
|
11, 12 |
1214 |
|
XM_011544368.2 |
XP_011542670.1
|
11, 12 |
1214 |
|
XM_011544369.2 |
XP_011542671.1
|
11, 12 |
1214 |
|
XM_011544370.2 |
XP_011542672.1
|
10, 11 |
992 |
|
XM_011544371.2 |
XP_011542673.1
|
4, 5 |
436 |
|
XM_017012974.1 |
XP_016868463.1
|
11, 12 |
1214 |
|
XM_017012975.1 |
XP_016868464.1
|
11, 12 |
1214 |
|
XM_017012976.1 |
XP_016868465.1
|
12, 13 |
1313 |
|
GenBank mRNA |
AF137334.1 |
- |
11, 12 |
1143 |
|
AF137335.1 |
- |
11, 12 |
1143 |
|
AJ242015.1 |
- |
11, 12 |
1151 |
|
AK225921.1 |
- |
|
1961 |
|
AK293382.1 |
- |
10, 11 |
993 |
|
AK303437.1 |
- |
11, 12 |
1247 |
|
BC136478.1 |
- |
11, 12 |
1130 |
9020467 |
View Full Product Details