Gene Transcripts
- Gene Symbol
- ADAM33
- Entrez Gene ID
- 80332
- Gene Name
- ADAM metallopeptidase domain 33
- Gene Aliases
- C20orf153, DJ964F7.1
- Chromosome Location
- Chr. 20: 3667973 - 3682131 on Build GRCh38
- UniGene ID
- Hs.173716
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282447.2 |
NP_001269376.1
|
4, 5 |
1021 |
|
NM_025220.4 |
NP_079496.1
|
4, 5 |
1021 |
|
NM_153202.3 |
NP_694882.1
|
4, 5 |
1021 |
|
XM_011529367.1 |
XP_011527669.1
|
4, 5 |
338 |
|
XM_011529373.1 |
XP_011527675.1
|
4, 5 |
375 |
|
XM_017028081.1 |
XP_016883570.1
|
4, 5 |
399 |
|
GenBank mRNA |
AB055891.1 |
- |
4, 5 |
422 |
|
AF466287.1 |
- |
4, 5 |
340 |
|
AK300429.1 |
- |
4, 5 |
389 |
|
AY358314.1 |
- |
4, 5 |
448 |
|
BC125113.1 |
- |
4, 5 |
382 |
40118831 |
DB225158.1 |
- |
4, 5 |
399 |
|
View Full Product Details