Gene Transcripts

Gene Symbol
SCGB2A2
Entrez Gene ID
4250
Gene Name
secretoglobin family 2A member 2
Gene Aliases
MGB1, UGB2
Chromosome Location
Chr. 11: 62270130 - 62273160 on Build GRCh38
UniGene ID
Hs.46452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002411.3 NP_002402.1 3 386
GenBank mRNA BG207094.1 - 1 85
U33147.1 - 3 384

View Full Product Details