Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002126.4 | NP_002117.1 | 4 | 2145 | |
XM_005257269.2 | XP_005257326.1 | 4 | 2128 | ||
XM_011524705.1 | XP_011523007.1 | 4 | 1602 | ||
XM_017024556.1 | XP_016880045.1 | 4 | 1599 | ||
GenBank mRNA | AK074289.1 | - | 3 | 1609 | |
BC036093.1 | - | 4 | 2099 | 5288156 | |
M95585.1 | - | 4 | 1942 |