Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004173.2 | NP_004164.2 | 3 | 1296 | |
GenBank mRNA | AJ000730.1 | - | 3 | 1309 | |
BC008814.2 | - | 3 | 1296 | 3958091 | |
BC062565.1 | - | 3 | 1290 | 5165369 | |
BC107160.1 | - | 2 | 1259 | 40033862 | |
BC107161.1 | - | 2 | 1259 | 40033864 | |
CR456580.1 | - | 2 | 1261 |