Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130107.1 | NP_001123579.1 | 3 | 754 | |
NM_005552.4 | NP_005543.2 | 3 | 754 | ||
NM_182923.3 | NP_891553.2 | 3 | 754 | ||
GenBank mRNA | AB451299.1 | - | 2 | 434 | |
AB451431.1 | - | 2 | 434 | ||
AF527736.1 | - | 3 | 704 | ||
AK294508.1 | - | 3 | 739 | ||
AY180164.2 | - | 3 | 701 | ||
BC008881.2 | - | 3 | 715 | 4301579 | |
L04733.1 | - | 3 | 710 |