Gene Transcripts

Gene Symbol
ARIH1
Entrez Gene ID
25820
Gene Name
ariadne RBR E3 ubiquitin protein ligase 1
Gene Aliases
ARI, HARI, HHARI, UBCH7BP
Chromosome Location
Chr. 15: 72474282 - 72586563 on Build GRCh38
UniGene ID
Hs.268787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005744.3 NP_005735.2 3 868
XM_011521438.2 XP_011519740.1 2 844
XM_011521439.2 XP_011519741.1 4 382
GenBank mRNA AB209432.1 - 4 497
AF072832.1 - 3 554
AJ130976.1 - 3 272
AJ243190.1 - 3 868
AK312715.1 - 3 867
BC051877.1 - 3 803 5742498

View Full Product Details