Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022341.1 | NP_071736.1 | 1153 | ||
GenBank mRNA | AF239156.1 | - | 1153 | ||
BC019912.1 | - | 1129 | 4994286 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032382.4 | NP_115758.3 | 2490 | ||
GenBank mRNA | BC017492.1 | - | 1294 |