Gene Transcripts

Gene Symbol
SULT1A2
Entrez Gene ID
6799
Gene Name
sulfotransferase family 1A member 2
Gene Aliases
HAST4, P-PST, ST1A2, STP2, TSPST2
Chromosome Location
Chr. 16: 28591943 - 28597079 on Build GRCh38
UniGene ID
Hs.546304
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001054.3 NP_001045.1 8 1026
NM_177528.2 NP_803564.1 8 1299
XM_006721077.3 XP_006721140.1 4 1563
XM_017023600.1 XP_016879089.1 8 1053
XM_017023601.1 XP_016879090.1 8 1481
XM_017023602.1 XP_016879091.1 9 1183
XM_017023603.1 XP_016879092.1 6 809
GenBank mRNA U28169.1 - 8 1021
U28170.1 - 8 1009
X78282.1 - 7 948

View Full Product Details