Gene Transcripts

Gene Symbol
MLLT11
Entrez Gene ID
10962
Gene Name
myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11
Gene Aliases
AF1Q
Chromosome Location
Chr. 1: 151059675 - 151068497 on Build GRCh38
UniGene ID
Hs.75823
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006818.3 NP_006809.1 2 1717
GenBank mRNA AK056089.1 - 2 1712
AK057755.1 - 3 950
BC006471.2 - 1 839 2823316
BC007079.1 - 2 981
BC008445.1 - 2 981 4103485
BC009624.1 - 2 997 3878959
BC021703.1 - 2 1055 4108071
BC022448.1 - 2 1989 4815214
BC032996.1 - 2 1014
U16954.1 - 2 1189

View Full Product Details