Gene Transcripts

Gene Symbol
C11orf71
Entrez Gene ID
54494
Gene Name
chromosome 11 open reading frame 71
Gene Aliases
URLC7
Chromosome Location
Chr. 11: 114391448 - 114400550 on Build GRCh38
UniGene ID
Hs.715083
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271562.1 NP_001258491.1 1 875
GenBank mRNA AB101209.1 - 1 864
AK000017.1 - 1 836
BC009041.1 - 1 804 3868744

View Full Product Details